Silicon(IV) nitride, alpha-phase, ca. 90%, Thermo Scientific Chemicals
Silicon(IV) nitride, alpha-phase, ca. 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silicon(IV) nitride, alpha-phase, ca. 90%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 042949.18
also known as 042949-18
Price (USD)/ Each
55.20
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
55.20
Add to cart
Silicon(IV) nitride, alpha-phase, ca. 90%, Thermo Scientific Chemicals
Catalog number042949.18
Price (USD)/ Each
55.20
-
Add to cart
Chemical Identifiers
CAS12033-89-5
IUPAC NameSilicon nitride
Molecular FormulaN4Si3
InChI KeyHQVNEWCFYHHQES-UHFFFAOYSA-N
SMILESN12[Si]34N5[Si]11N3[Si]25N41
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to grey or brown
FormPowder
Assay from Suppliers CofAAlpha phase: ≥87.0%
Particle Size≤10µm
Silicon(IV) nitride is used in the automobile industry as a material for engine parts such as diesel engines and glowplugs. It is used for rocker arm pads for lower wear in spark-ignition engines. Ceramics made from this show good shock resistance. Similarly bearings made from this find application in the main engines of NASA’s space shuttle. It acts as an insulator and in the manufacture of integrated circuits.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only