Thermo Scientific Chemicals

Borane-dimethyl sulfide complex, 2M in toluene, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 042963.AE
100 mL, Each
Thermo Scientific Chemicals

Borane-dimethyl sulfide complex, 2M in toluene, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 042963.AE
100 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 042963.AE
also known as 042963-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13292-87-0
IUPAC Name
(methylsulfanyl)methane borane
Molecular Formula
C2H9BS
InChI Key
RMHDLBZYPISZOI-UHFFFAOYSA-N
SMILES
B.CSC
Form
Liquid
Appearance (Color)
Colorless
Assay (unspecified)
1.90-2.20M ( gas evolution)

Description

Borane-dimethyl sulfide complex is used for hydroborations and reductions. Used with a dendrimeric supported L-pyrrolidinol in the asymmetric reduction of indanones and tetralones. CBS-catalyzed asymmetric reduction of ferrocenyl-1,3-diketones to 1,3-diols. Highly enantioselective reduction of ketones catalyzed by C3-symmetric tripodal hydroxyamides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text