Thermo Scientific Chemicals

Borane-dimethyl sulfide complex, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 042967.AA
5 mL, Each
Thermo Scientific Chemicals

Borane-dimethyl sulfide complex, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 042967.AA
5 mL, Each
Quantity
Catalog number: 042967.AA
also known as 042967-AA
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13292-87-0
IUPAC Name
(methylsulfanyl)methane borane
Molecular Formula
C2H9BS
InChI Key
RMHDLBZYPISZOI-UHFFFAOYSA-N
SMILES
B.CSC
Assay from Supplier's CofA
≥93.0 to ≤98.0%
Appearance (Color)
Colorless to pale yellow
Form
Liquid

Description

Borane- dimethyl sulfide is used for hydroboration and reduction reactions. It is used in the reduction of aldehydes, ketones, epoxides and carboxylic acids to give corresponding alcohols. It is also involved in the Corey-Itsuno reduction. It acts as an intermediate in the preparation of highly pure semiconductor. It is also employed as a rocket propellant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text