Borane-dimethyl sulfide complex, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Borane-dimethyl sulfide complex, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Borane-dimethyl sulfide complex, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
100 mL
500 mL
Catalog number 042967.AC
also known as 042967-AC
Price (USD)/ Each
47.65
Online exclusive
53.10 
Save 5.45 (10%)
-
Add to cart
Quantity:
25 mL
Request bulk or custom format
Price (USD)/ Each
47.65
Online exclusive
53.10 
Save 5.45 (10%)
Add to cart
Borane-dimethyl sulfide complex, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Catalog number042967.AC
Price (USD)/ Each
47.65
Online exclusive
53.10 
Save 5.45 (10%)
-
Add to cart
Chemical Identifiers
CAS13292-87-0
IUPAC Name(methylsulfanyl)methane borane
Molecular FormulaC2H9BS
InChI KeyRMHDLBZYPISZOI-UHFFFAOYSA-N
SMILESB.CSC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥93.0 to ≤98.0%
Appearance (Color)Colorless to pale yellow
FormLiquid
Borane- dimethyl sulfide is used for hydroboration and reduction reactions. It is used in the reduction of aldehydes, ketones, epoxides and carboxylic acids to give corresponding alcohols. It is also involved in the Corey-Itsuno reduction. It acts as an intermediate in the preparation of highly pure semiconductor. It is also employed as a rocket propellant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only