Thermo Scientific Chemicals

Boron carbide, Thermo Scientific Chemicals

Catalog number: 043002.36
500 g, Each
Thermo Scientific Chemicals

Boron carbide, Thermo Scientific Chemicals

Catalog number: 043002.36
500 g, Each
Quantity
Catalog number: 043002.36
also known as 043002-36
Price (USD)
Have Questions?

Chemical Identifiers

CAS
12069-32-8
IUPAC Name
2,3,4,5-tetraboratetracyclo[2.1.0.0¹,³.0²,⁵]pentane
Molecular Formula
CB4
InChI Key
INAHAJYZKVIDIZ-UHFFFAOYSA-N
SMILES
B12B3B4B1C234
Appearance (Color)
Black
Form
Powder
Assay from Suppliers CofA
Report: % Boron Carbide (B4C)
Particle Size
<10 microns (APS)

Description

Boron carbide is a hard boron-carbon ceramic material used in tank armor, bulletproof vests, engine sabotage powders, neutron absorber, cutting tools and dies and in brake linings of vehicles. It acts as an antioxidant additives in magnesia-carbon bricks. It is used as a precursor in the production of boron containing materials such as titanium boride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text