Thermo Scientific Chemicals

Platinum Iridium wire, 0.127mm (0.005in) dia, annealed, 99.9% (metals basis excluding precious metals), Thermo Scientific Chemicals

Catalog number: 043019.BW
50 cm, Each
Thermo Scientific Chemicals

Platinum Iridium wire, 0.127mm (0.005in) dia, annealed, 99.9% (metals basis excluding precious metals), Thermo Scientific Chemicals

Catalog number: 043019.BW
50 cm, Each
Quantity
Catalog number: 043019.BW
also known as 043019-BW
Price (USD)
Quantity
-
Have Questions?
Melting Point
1775?C
Form
Wire, 0.2mm (0.008in) diameter, annealed.
Assay from Supplier's CofA
Pt:Ir; 90:10 wt%
Appearance (Color)
Silver gray
Assay from Supplier's CofA
≥99.9% (metals basis excluding precious metals)

Description

Vacuum evaporation deposition, scanning and tunneling microscopy, corrosion resistant electrodes.Platinum-iridium wire is used in platinum jewellery, particularly watch cases. They have been used for spinnerets in the manufacture of synthetic fibres, in metrology and alloy is fabrication of metal microelectrodes. It is used in electrical stimulation of nervous tissue and electrophysiological recordings. Pt/Ir alloy has been used in mechanical and electrochemical devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Vacuum evaporation deposition, scanning and tunneling microscopy, corrosion resistant electrodes
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text