Aluminum powder, -100+325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Aluminum powder, -100+325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum powder, -100+325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 043022.30
also known as 043022-30
Price (USD)/ Each
75.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
89.00 
Save 13.35 (15%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
75.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
89.00 
Save 13.35 (15%)
Add to cart
Aluminum powder, -100+325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Catalog number043022.30
Price (USD)/ Each
75.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
89.00 
Save 13.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤0.5%
FormPowder
Particle Size-100+325 mesh (between 44 to 149 microns): Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only