Thermo Scientific Chemicals

Titanium sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 043043.KS
1 Ea., Each
Thermo Scientific Chemicals

Titanium sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 043043.KS
1 Ea., Each
Quantity
Catalog number: 043043.KS
also known as 043043-KS
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7440-32-6
IUPAC Name
titanium
Molecular Formula
Ti
InChI Key
RTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES
[Ti]
Assay from Supplier's CofA
≥99.995% (metals basis)
Appearance (Color)
Gray
Form
Sputtering target, 50.8mm (2.0in) diameter x 6.35mm (0.250in) thick.

Description

Titanium sputtering target and its alloys are characterized by their lightness, strength and corrosion resistance and are used widely in aerospace applications. In addition, these properties also make the material suitable for medical applications (e.g. replacement hip joints). Titanium dioxide is used as a white pigment in paints and plastics as it provides great opacity. The same material is also used in the manufacture of heat resisting and durable glass, the TiO2 replacing certain proportions of the soda. Titanium carbide is used to manufacture cemented carbides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text