Thermo Scientific Chemicals

Titanium(IV) oxide, rutile, 99.5% min (metals basis), Thermo Scientific Chemicals

Catalog number: 043047.30
250 g, Each
Thermo Scientific Chemicals

Titanium(IV) oxide, rutile, 99.5% min (metals basis), Thermo Scientific Chemicals

Catalog number: 043047.30
250 g, Each
Quantity
Catalog number: 043047.30
also known as 043047-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1317-80-2
Total Metal Impurities
0.5% max.

Description

Titanium dioxide is used in paints, plastics, paper, and foods. It is used as a UV-resistant material. It shows useful applications in earth sciences. It is also useful in quantitative provenance studies. It is also useful for the photocatalytic applications. It is also useful in high temperature superconducting resonators. It is also used as a material in the memristor, a new electronic circuit element. It can be used for solar energy conversion based on dye, to cleave protein that contains the amino acid proline polymer, quantum dot sensitized nanocrystalline TiO2 solar cells using conjugated polymers as solid electrolytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text