Thermo Scientific Chemicals

Iridium(IV) oxide dihydrate, Premion™, 99.99% (metals basis), Ir 73% min, Thermo Scientific Chemicals

Catalog number: 043051.03
1 g, Each
Thermo Scientific Chemicals

Iridium(IV) oxide dihydrate, Premion™, 99.99% (metals basis), Ir 73% min, Thermo Scientific Chemicals

Catalog number: 043051.03
1 g, Each
Quantity
Catalog number: 043051.03
also known as 043051-03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
30980-84-8
IUPAC Name
dioxoiridium hydrate
Molecular Formula
H2IrO3
InChI Key
YEPXCFHWKVNVAX-UHFFFAOYSA-N
SMILES
O.O=[Ir]=O
Total Metal Impurities
0.01% max.
Assay (unspecified)
Ir 73% min

Description

Iridium(IV) oxide dihydrate is used with other rare oxides in the coating of anode-electrodes for industrial electrolysis and in microelectrodes for electrophysiology research. It is also used for reference electrode, which finds application in various fields biology, food industry, nuclear field, oil and gas industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text