Germanium(IV) ethoxide, 99.995% (metals basis), Thermo Scientific Chemicals
Germanium(IV) ethoxide, 99.995% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Germanium(IV) ethoxide, 99.995% (metals basis), Thermo Scientific Chemicals

Preparation of GeO2 by CVD; intermediate in sol-gel synthesis
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 043052.03
also known as 043052-03
Price (USD)/ Each
63.40
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
63.40
Add to cart
Germanium(IV) ethoxide, 99.995% (metals basis), Thermo Scientific Chemicals
Catalog number043052.03
Price (USD)/ Each
63.40
-
Add to cart
Specifications
Chemical Name or MaterialGermanium (IV) ethoxide
CAS14165-55-0
Health Hazard 1H226-H301+H311+H331-H315-H319-H335
Health Hazard 2GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P363-P370+P378q-P501c
View more
Preparation of Germanium oxide by CVD.Sol-gel intermediate for optical fibers and photosensitive SiO2-BeO2 glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only