Thermo Scientific Chemicals

Toluene, Environmental Grade, 99.8+%, Thermo Scientific Chemicals

Catalog number: 043061.K7
4 L, Each
Thermo Scientific Chemicals

Toluene, Environmental Grade, 99.8+%, Thermo Scientific Chemicals

Catalog number: 043061.K7
4 L, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 043061.K7
also known as 043061-K7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-88-3
IUPAC Name
toluene
Molecular Formula
C7H8
InChI Key
YXFVVABEGXRONW-UHFFFAOYSA-N
SMILES
CC1=CC=CC=C1
Comment
Halogenated residueby GC-ECD (as Heptachlorepoxide) 5ng/L
Residual water
0.03%
Color (APHA)
10

Description

Suitable for pesticide residue analysisToluene is used as a solvent for paints, paint thinners, silicone sealants, many chemical reactants, rubber, printing ink, adhesives (glues), lacquers, leather tanners and disinfectants. It is an important raw material to synthesize pharmaceuticals, paints, resins, dyes, toluene diisocyanate and explosives like trinitrotoluene. It is used as a standard material for chromatography and analytical reagent. It plays an important role in the manufacture of benzene, xylene, phenol, poly urethane polymers, cosmetics and jet fuel blends. It is used as fuel additive to increase octane ratings and as a coolant for its good heat transfer capabilities in sodium cold traps used in nuclear reactor system loops.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Maximum level of impurities: Color (APHA) 10, Halogenated residue by GC-ECD, (as Heptachlorepoxide) 5ng/L, Water 0.03%
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text