Poly(vinylbenzyl chloride), Thermo Scientific Chemicals
Poly(vinylbenzyl chloride), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Poly(vinylbenzyl chloride), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
50 g
Catalog number 043067.09
also known as 043067-09
Price (USD)/ Each
157.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
157.00
Add to cart
Poly(vinylbenzyl chloride), Thermo Scientific Chemicals
Catalog number043067.09
Price (USD)/ Each
157.00
-
Add to cart
Chemical Identifiers
CAS9080-67-5
SpecificationsSpecification SheetSpecification Sheet
Molecular Weight25,000 - 50,000
In scientific and educational research. In synthesis of polymer supported reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only