Calcium carbonate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals
Calcium carbonate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium carbonate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
500 g
Catalog number 043073.14
Price (USD)/ Each
198.65
Online exclusive
220.00 
Save 21.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
198.65
Online exclusive
220.00 
Save 21.35 (10%)
Add to cart
Calcium carbonate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals
Catalog number043073.14
Price (USD)/ Each
198.65
Online exclusive
220.00 
Save 21.35 (10%)
-
Add to cart
Chemical Identifiers
CAS471-34-1
IUPAC Namecalcium carbonate
Molecular FormulaCCaO3
InChI KeyVTYYLEPIZMXCLO-UHFFFAOYSA-L
SMILES[Ca++].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.003% max.
Calcium carbonate is used in medicine and agriculture. It is also used in flue gas desulfurization in fossil fuel power stations, production of toothpaste, ceramics/glazing applications, purification of iron from iron ore, and to clean tarnish on silver. It is used in the refining of sugar, as filler in thermosetting resins, as an abrasive, as an extender in paints, and as a formation-bridging filtercake-sealing agent in oil industry. It is used in swimming pools for controlling the pH, to improve the rigidity of polypropylene compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only