Assay from Supplier's CofAFurther Ingredients: Ethylenediamine tetraacetic acid (EDTA) (Reagent Grade) CAS 60-00-4 EEC No. 200-449-4
Assay from Supplier's CofAFurther Ingredients: Sodium hydroxide (Reagent Grade) CAS 1310-73-2 EEC No. 215-185-5
Appearance (Color)Clear to pale yellow
Assay from Supplier's CofAAssay: N/A
Assay from Supplier's CofAFurther Ingredients: Methyl alcohol (Reagent Grade) CAS 67-56-1 EEC No. 200-659-6
View more