Thermo Scientific Chemicals

Ionic strength adjuster solution for chloride or sulfide electrodes

Catalog number: 043075.AP
500 mL, Each
Thermo Scientific Chemicals

Ionic strength adjuster solution for chloride or sulfide electrodes

Catalog number: 043075.AP
500 mL, Each
Quantity
Catalog number: 043075.AP
also known as 043075-AP
Price (USD)
Quantity
-
Have Questions?
Form
Liquid
Assay from Supplier's CofA
5.00M ? 0.25M

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text