Further Ingredients: Ammonium hydroxide (ACS grade) 1336-21-6, EEC No 215-647-6
Form
Liquid
Assay from Supplier's CofA
Further Ingredients: Ammonium chloride (ACS grade) CAS 12125-02-9, EEC No 235-186-4
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.