Thermo Scientific Chemicals

Sulfuric acid, 3.0N Standardized Solution, Thermo Scientific Chemicals

Catalog number: 043082.K2
1 L, Each
Thermo Scientific Chemicals

Sulfuric acid, 3.0N Standardized Solution, Thermo Scientific Chemicals

Catalog number: 043082.K2
1 L, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 043082.K2
also known as 043082-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7664-93-9
IUPAC Name
sulfuric acid
Molecular Formula
H2O4S
InChI Key
QAOWNCQODCNURD-UHFFFAOYSA-N
SMILES
OS(O)(=O)=O
Appearance (Color)
Colorless to pale yellow
Assay from Supplier's CofA
2.994 N - 3.006 N
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text