Graphite foil, 1mm (0.04in) thick, 97% (metals basis), Thermo Scientific Chemicals
Graphite foil, 1mm (0.04in) thick, 97% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Graphite foil, 1mm (0.04in) thick, 97% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
30 x 500 cm
30 x 30 cm
30 x 100 cm
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 043083.RX
Price (USD)/ Each
-
Quantity:
30 x 500 cm
Request bulk or custom format
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialGraphite foil
Name Note1mm (0.04 in.) thick
Recommended StorageAmbient temperatures
EINECS Number231-955-3
TSCAYes
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only