Silver nitrate, Premion™, 99.995% (metals basis), Ag 63% min, Thermo Scientific Chemicals
Silver nitrate, Premion™, 99.995% (metals basis), Ag 63% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver nitrate, Premion™, 99.995% (metals basis), Ag 63% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 043087.06
also known as 043087-06
Price (USD)/ Each
39.65
Online exclusive
43.50 
Save 3.85 (9%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
39.65
Online exclusive
43.50 
Save 3.85 (9%)
Add to cart
Silver nitrate, Premion™, 99.995% (metals basis), Ag 63% min, Thermo Scientific Chemicals
Catalog number043087.06
Price (USD)/ Each
39.65
Online exclusive
43.50 
Save 3.85 (9%)
-
Add to cart
Chemical Identifiers
CAS7761-88-8
IUPAC Namesilver(1+) nitrate
Molecular FormulaAgNO3
InChI KeySQGYOTSLMSWVJD-UHFFFAOYSA-N
SMILES[Ag+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Ag 63% min
Total Metal Impurities0.005% max.
Silver nitrate is employed in Silver plating, photography, manufacturing of other silver compounds, mirrors, coloring porcelain, and staining in histology. It finds use as a stain in scanning electron microscopy, and a key ingredient in several silver-based antiseptic and antibacterial pharmaceutical compositions. Silver(I) nitrate can be used to separate mixtures of alkenes by selective absorption, and for deprotection and oxidation reactions in organic synthesis. It is widely used as an analytical reagent owing to its immediate reaction with halide ions forming insoluble precipitate of silver(I) halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only