Platinum(II) chloride, Premion™, 99.99+% (metals basis), Pt 73% min, Thermo Scientific Chemicals
Platinum(II) chloride, Premion™, 99.99+% (metals basis), Pt 73% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Platinum(II) chloride, Premion™, 99.99+% (metals basis), Pt 73% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 043090.03
also known as 043090-03
Price (USD)/ Each
266.65
Online exclusive
296.00 
Save 29.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
266.65
Online exclusive
296.00 
Save 29.35 (10%)
Add to cart
Platinum(II) chloride, Premion™, 99.99+% (metals basis), Pt 73% min, Thermo Scientific Chemicals
Catalog number043090.03
Price (USD)/ Each
266.65
Online exclusive
296.00 
Save 29.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10025-65-7
IUPAC Nameplatinum(2+) dichloride
Molecular FormulaCl2Pt
InChI KeyCLSUSRZJUQMOHH-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Pt++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Pt 73% min
Total Metal Impurities0.01% max.
Platinum(II) chloride is used as a catalyst for a variety of C-C bond, C-O bond and C-N bond formation reactions. It acts as a precursor to prepare other platinum compounds such as cisplatin and dichloro(cycloocta-1,5-diene)platinum(II).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only