Thermo Scientific Chemicals

Platinum(II) chloride, Premion™, 99.99+% (metals basis), Pt 73% min, Thermo Scientific Chemicals

Catalog number: 043090.06
5 g, Each
Thermo Scientific Chemicals

Platinum(II) chloride, Premion™, 99.99+% (metals basis), Pt 73% min, Thermo Scientific Chemicals

Catalog number: 043090.06
5 g, Each
Quantity
Catalog number: 043090.06
also known as 043090-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10025-65-7
IUPAC Name
platinum(2+) dichloride
Molecular Formula
Cl2Pt
InChI Key
CLSUSRZJUQMOHH-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Pt++]
Assay (unspecified)
Pt 73% min
Total Metal Impurities
0.01% max.

Description

Platinum(II) chloride is used as a catalyst for a variety of C-C bond, C-O bond and C-N bond formation reactions. It acts as a precursor to prepare other platinum compounds such as cisplatin and dichloro(cycloocta-1,5-diene)platinum(II).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text