Thermo Scientific Chemicals

Poly(Allylamine Hydrochloride), Thermo Scientific Chemicals

Catalog number: 043092.18
50 g, Each
Thermo Scientific Chemicals

Poly(Allylamine Hydrochloride), Thermo Scientific Chemicals

Catalog number: 043092.18
50 g, Each
Quantity
Catalog number: 043092.18
also known as 043092-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
71550-12-4
Appearance (Color)
Cream to yellow
Form
Powder or crystals or crystalline powder
pH
Report
Solution Test
Solubility test: Report
Water Content
≤5.0%

Description

Poly(allylamine hydrochloride) is used in film and polymer industries and cell encapsulation. Further, it is used to make redox hydrogel- modified electrodes for measuring enzyme responses. In addition, it is used in the preparation of an antireflective coating consists of alternate layers of poly(allyamine hydrochloride) and silica nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text