Thermo Scientific Chemicals

Ruthenium(IV) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 043098.06
5 g, Each
Thermo Scientific Chemicals

Ruthenium(IV) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 043098.06
5 g, Each
Quantity
Catalog number: 043098.06
also known as 043098-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12166-20-0
IUPAC Name
disulfanylideneruthenium
Molecular Formula
RuS2
InChI Key
XNQLQKWNBPVVGB-UHFFFAOYSA-N
SMILES
S=[Ru]=S
Total Metal Impurities
0.1% max.

Description

Ruthenium(IV) sulfide is used to prepare sintered material, which is a semiconductor. It acts as an electrode and posses high catalytic activity for oxygen evolution from aqueous solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text