Silicon(IV) oxide, 50% in H{2}O, colloidal dispersion, Thermo Scientific Chemicals
Silicon(IV) oxide, 50% in H{2}O, colloidal dispersion, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silicon(IV) oxide, 50% in H{2}O, colloidal dispersion, Thermo Scientific Chemicals

S.A. 180m2/g
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Catalog number 043109.A1
also known as 043109-A1
Price (USD)/ Each
29.70
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
29.70
Add to cart
Silicon(IV) oxide, 50% in H{2}O, colloidal dispersion, Thermo Scientific Chemicals
Catalog number043109.A1
Price (USD)/ Each
29.70
-
Add to cart
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Surface Area180m ²/g
Particle Size0.02 micron ? 0.002
Concentration (weight/weight)50% min
Silica is used primarily in the production of glass for windows, drinking glasses, beverage bottles, and many other uses. The majority of optical fibers for telecommunication are also made from silica. It is a primary raw material for many ceramics such as earthenware, stoneware, and porcelain. The main ingredient in sand casting for the manufacture of a large number of metallic components in engineering and other applications. Primarily as a flow agent in powdered foods, or to adsorb water in hygroscopic applications. It is the primary component of diatomaceous earth. Colloidal silica is also used as a wine, beer, and juice fining agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only