Thermo Scientific Chemicals

Silicon(IV) oxide, 50% in H{2}O, colloidal dispersion, Thermo Scientific Chemicals

Catalog number: 043109.A7
5 kg, Each
Thermo Scientific Chemicals

Silicon(IV) oxide, 50% in H{2}O, colloidal dispersion, Thermo Scientific Chemicals

Catalog number: 043109.A7
5 kg, Each
Quantity
Catalog number: 043109.A7
also known as 043109-A7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7631-86-9
IUPAC Name
silanedione
Molecular Formula
O2Si
InChI Key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES
O=[Si]=O
Surface Area
180m ²/g
Particle Size
0.02 micron ? 0.002
Concentration (weight/weight)
50% min

Description

Silica is used primarily in the production of glass for windows, drinking glasses, beverage bottles, and many other uses. The majority of optical fibers for telecommunication are also made from silica. It is a primary raw material for many ceramics such as earthenware, stoneware, and porcelain. The main ingredient in sand casting for the manufacture of a large number of metallic components in engineering and other applications. Primarily as a flow agent in powdered foods, or to adsorb water in hygroscopic applications. It is the primary component of diatomaceous earth. Colloidal silica is also used as a wine, beer, and juice fining agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text