Aluminum oxide, Aerosol Refractory Brushable Paint, Thermo Scientific Chemicals
Aluminum oxide, Aerosol Refractory Brushable Paint, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum oxide, Aerosol Refractory Brushable Paint, Thermo Scientific Chemicals

Provides protective high temperature Al2O3 coating on ceramic, graphite, or metal
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number 043119.A1
also known as 043119-A1
Price (USD)/ Each
326.65
Online exclusive
363.00 
Save 36.35 (10%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
326.65
Online exclusive
363.00 
Save 36.35 (10%)
Add to cart
Aluminum oxide, Aerosol Refractory Brushable Paint, Thermo Scientific Chemicals
Catalog number043119.A1
Price (USD)/ Each
326.65
Online exclusive
363.00 
Save 36.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialAluminum oxide
Name NoteAerosol Refractory Brushable Paint
CAS1344-28-1
ColorWhite
Health Hazard 1H336
View more
Provides protective high temperature Al{2}O{3} coating on ceramic, graphite, or metalAluminum oxide, Aerosol Refractory Brushable Paint is used as barrier layer between dissimilar materials to prevent reaction and interactions during high temperature. It also acts as a solvent-based paint. It also provides protective high temperature alumina coating on ceramic, graphite and metal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
For Manufacturing and Laboratory Use. Not for beverage use