Lanthanum(III) chloride hydrate, 99.9% (REO), Thermo Scientific Chemicals
Lanthanum(III) chloride hydrate, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lanthanum(III) chloride hydrate, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number 043120.36
also known as 043120-36
Price (USD)/ Each
203.65
Online exclusive
226.00 
Save 22.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
203.65
Online exclusive
226.00 
Save 22.35 (10%)
Add to cart
Lanthanum(III) chloride hydrate, 99.9% (REO), Thermo Scientific Chemicals
Catalog number043120.36
Price (USD)/ Each
203.65
Online exclusive
226.00 
Save 22.35 (10%)
-
Add to cart
Chemical Identifiers
CAS20211-76-1
IUPAC Namelanthanum(3+) trichloride
Molecular FormulaCl3La
InChI KeyICAKDTKJOYSXGC-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[La+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Lanthanum(III) chloride is useful as precursor for synthesis of aqueous sol-gel lanthanum phosphate nano rods,as a filter aid,an effective flocculent and precipitation phosphate. It is also used as a catalyst for the high pressure oxidative chlorination of methane to chloromethane and organic synthesis it acts as Lewis acid catalyst. It is used as a scintillator material and in Gamma Detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only