Lanthanum(III) oxalate hydrate, 99.9% (REO), Thermo Scientific Chemicals
Lanthanum(III) oxalate hydrate, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lanthanum(III) oxalate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 043122.22
also known as 043122-22
Price (USD)/ Each
53.70
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
53.70
Add to cart
Lanthanum(III) oxalate hydrate, 99.9% (REO), Thermo Scientific Chemicals
Catalog number043122.22
Price (USD)/ Each
53.70
-
Add to cart
Chemical Identifiers
CAS79079-18-8
IUPAC Namedilanthanum(3+) trioxalate
Molecular FormulaC6La2O12
InChI KeyOXHNIMPTBAKYRS-UHFFFAOYSA-H
SMILES[La+3].[La+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Lanthanum Oxalate is mainly applied in specialty glass, phosphors, water treatment and catalyst. This material use in glass, ceramics, paper, textile, coating, fine and specialty chemicals. In petroleum cracking catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only