Thermo Scientific Chemicals

Methacrolein, 96%, stab. with hydroquinone, Thermo Scientific Chemicals

Catalog number: 043124.AA
5 mL, Each
Thermo Scientific Chemicals

Methacrolein, 96%, stab. with hydroquinone, Thermo Scientific Chemicals

Catalog number: 043124.AA
5 mL, Each
Quantity
Catalog number: 043124.AA
also known as 043124-AA
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
78-85-3
IUPAC Name
2-methylprop-2-enal
Molecular Formula
C4H6O
InChI Key
STNJBCKSHOAVAJ-UHFFFAOYSA-N
SMILES
CC(=C)C=O
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay from Supplier's CofA
≥95.0%
Comment
Stabilized with hydroquinone

Description

Methacrolein is used in the manufacturing of polymers, synthetic resins and plastics. It acts as a raw material of methylmalonic acid, thermoplastic fuel and in the preparation of 2,3-dibromo-2-methyl-propionaldehyde. It is also used to study the effect limonene oxidation products and the terpene oxidation product.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text