Methacrolein, 96%, stab. with hydroquinone, Thermo Scientific Chemicals
Methacrolein, 96%, stab. with hydroquinone, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methacrolein, 96%, stab. with hydroquinone, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
Catalog number 043124.AC
Price (USD)/ Each
190.65
Online exclusive
212.00 
Save 21.35 (10%)
-
Add to cart
Quantity:
25 mL
Request bulk or custom format
Price (USD)/ Each
190.65
Online exclusive
212.00 
Save 21.35 (10%)
Add to cart
Methacrolein, 96%, stab. with hydroquinone, Thermo Scientific Chemicals
Catalog number043124.AC
Price (USD)/ Each
190.65
Online exclusive
212.00 
Save 21.35 (10%)
-
Add to cart
Chemical Identifiers
CAS78-85-3
IUPAC Name2-methylprop-2-enal
Molecular FormulaC4H6O
InChI KeySTNJBCKSHOAVAJ-UHFFFAOYSA-N
SMILESCC(=C)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay from Supplier's CofA≥95.0%
CommentStabilized with hydroquinone
Methacrolein is used in the manufacturing of polymers, synthetic resins and plastics. It acts as a raw material of methylmalonic acid, thermoplastic fuel and in the preparation of 2,3-dibromo-2-methyl-propionaldehyde. It is also used to study the effect limonene oxidation products and the terpene oxidation product.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only