Nickel wire, 0.25mm (0.01in) dia, annealed, 99.98% (metals basis), Thermo Scientific Chemicals
Nickel wire, 0.25mm (0.01in) dia, annealed, 99.98% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel wire, 0.25mm (0.01in) dia, annealed, 99.98% (metals basis), Thermo Scientific Chemicals

Form: Approx.0.44g/m, Temper: soft
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 m
5 m
Catalog number 043131.G9
also known as 043131-G9
Price (USD)/ Each
178.65
Online exclusive
198.00 
Save 19.35 (10%)
-
Add to cart
Quantity:
25 m
Request bulk or custom format
Price (USD)/ Each
178.65
Online exclusive
198.00 
Save 19.35 (10%)
Add to cart
Nickel wire, 0.25mm (0.01in) dia, annealed, 99.98% (metals basis), Thermo Scientific Chemicals
Catalog number043131.G9
Price (USD)/ Each
178.65
Online exclusive
198.00 
Save 19.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery
Assay from Supplier's CofA≥99.98% (metals basis)
FormWire, 0.25mm (0.01in) diameter, annealed.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only