Thermo Scientific Chemicals

2,3-Dimethylbutane, 98+%, Thermo Scientific Chemicals

Catalog number: 043142.AK
250 mL, Each
Thermo Scientific Chemicals

2,3-Dimethylbutane, 98+%, Thermo Scientific Chemicals

Catalog number: 043142.AK
250 mL, Each
Quantity
Catalog number: 043142.AK
also known as 043142-AK
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79-29-8
IUPAC Name
2,3-dimethylbutane
Molecular Formula
C6H14
InChI Key
ZFFMLCVRJBZUDZ-UHFFFAOYSA-N
SMILES
CC(C)C(C)C
Appearance (Color)
Colorless
Assay (GC)
≥98.0%
Form
Liquid

Description

2,3-Dimethylbutane is used in high octane fuels and in organic synthesis. It is also used as a gas chromatography standard.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text