2,3-Dimethylbutane, 98+%, Thermo Scientific Chemicals
2,3-Dimethylbutane, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,3-Dimethylbutane, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Catalog number 043142.K2
also known as 043142-K2
Price (USD)/ Each
680.00
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
680.00
Add to cart
2,3-Dimethylbutane, 98+%, Thermo Scientific Chemicals
Catalog number043142.K2
Price (USD)/ Each
680.00
-
Add to cart
Chemical Identifiers
CAS79-29-8
IUPAC Name2,3-dimethylbutane
Molecular FormulaC6H14
InChI KeyZFFMLCVRJBZUDZ-UHFFFAOYSA-N
SMILESCC(C)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Assay (GC)≥98.0%
FormLiquid
2,3-Dimethylbutane is used in high octane fuels and in organic synthesis. It is also used as a gas chromatography standard.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only