5-Sulfosalicylic acid dihydrate, ACS, 99+%, Thermo Scientific Chemicals
5-Sulfosalicylic acid dihydrate, ACS, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Sulfosalicylic acid dihydrate, ACS, 99+%, Thermo Scientific Chemicals

Reagent for precipitation of proteins
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number 043144.22
also known as 043144-22
Price (USD)/ Each
55.65
Online exclusive
62.20 
Save 6.55 (11%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
55.65
Online exclusive
62.20 
Save 6.55 (11%)
Add to cart
5-Sulfosalicylic acid dihydrate, ACS, 99+%, Thermo Scientific Chemicals
Catalog number043144.22
Price (USD)/ Each
55.65
Online exclusive
62.20 
Save 6.55 (11%)
-
Add to cart
Chemical Identifiers
CAS5965-83-3
IUPAC Name2-hydroxy-5-sulfobenzoic acid; bis(methane)
Molecular FormulaC9H14O6S
InChI KeyNFYHZVWMQHQKRU-UHFFFAOYSA-N
SMILESC.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)99.0 - 101.0% ( as a Dihydrate)
Formsolid
Chloride content0.001%, max
Impurity contentSalicylic acid: 0.04%, max
View more
Reagent for precipitation of proteins5-Sulfosalicylic acid dihydrate is used as a metal scavenger and it forms proton-transfer dye complexes with diazo compounds such as 4-(phenyldiazenyl)aniline. It is employed as a preparative measure for removing proteins prior to chromatographic analysis. It plays a vital role in reducing and fixation of proteins in agarose and polyacrylamide gels. It acts as a surfactant and lubricating grease additive. It is also used in urine tests to determine urine protein content. Further, it acts as an intermediate in the synthesis of active pharmaceutical ingredients like doxycycline and methacycline. In addition to this, it is used as a spray reagent for the detection of amino acids on thin layer chromatography (TLC) plates and as a doping agent for polyaniline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only