Thermo Scientific Chemicals

Methyl cellulose, viscosity 400 cPs

Catalog number: 043147.22
100 g, Each
Thermo Scientific Chemicals

Methyl cellulose, viscosity 400 cPs

Catalog number: 043147.22
100 g, Each
Quantity
Catalog number: 043147.22
also known as 043147-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9004-67-5
IUPAC Name
3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane
Molecular Formula
C20H38O11
InChI Key
YLGXILFCIXHCMC-UHFFFAOYNA-N
SMILES
COCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC
Form
Powder
Viscosity
300-560 mPas/cPs (2% solution, 20°C)
Appearance (Color)
White to grey
Residue on ignition
≤2.0%
Assay from Suppliers CofA
Assay, dried basis (methoxy groups): 26.0-33.0%

Description

Methyl cellulose is used as a thickener and emulsifier in various food and cosmetic products like hair shampoos, toothpastes and liquid soaps. It is used in the preparation of capsules in nutritional supplements. It is also used as a gel in advanced cookery and as a lubricant. It is also involved in the treatment of constipation. It acts as a buffer additive in capillary electrophoresis to control electoosmotic flow for improved separations. Further, it is used as a performance additive in construction materials and as sizing in the production of papers and textiles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text