Thermo Scientific Chemicals

4-Methyl-2-pentanone, HPLC Grade, 99+%, Thermo Scientific Chemicals

Catalog number: 043170.M1
4 x 1 L, Each
Thermo Scientific Chemicals

4-Methyl-2-pentanone, HPLC Grade, 99+%, Thermo Scientific Chemicals

Catalog number: 043170.M1
4 x 1 L, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 043170.M1
also known as 043170-M1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-10-1
IUPAC Name
4-methylpentan-2-one
Molecular Formula
C6H12O
InChI Key
NTIZESTWPVYFNL-UHFFFAOYSA-N
SMILES
CC(C)CC(C)=O
Color (APHA)
10
Residue on Evaporation
5ppm
Residual water
0.1%

Description

4-Methyl-2-pentanone is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text