Thermo Scientific Chemicals

Triethylgermanium hydride, 97%, Thermo Scientific Chemicals

Catalog number: 043176.06
5 g, Each
Thermo Scientific Chemicals

Triethylgermanium hydride, 97%, Thermo Scientific Chemicals

Catalog number: 043176.06
5 g, Each
Quantity
Catalog number: 043176.06
also known as 043176-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1188-14-3
IUPAC Name
triethylgermane
Molecular Formula
C6H16Ge
InChI Key
LTLGQZPDSQYFSB-UHFFFAOYSA-N
SMILES
CC[GeH](CC)CC
Form
Liquid
Assay from Supplier's CofA
≥96.0%

Description

Co-reactant for the preparation of germanium acrylates. They are the Synthetic Reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text