Thermo Scientific Chemicals

Ammonium hexafluorogermanate, 99.99% min (metals basis), Thermo Scientific Chemicals

Catalog number: 043179.14
25 g, Each
Thermo Scientific Chemicals

Ammonium hexafluorogermanate, 99.99% min (metals basis), Thermo Scientific Chemicals

Catalog number: 043179.14
25 g, Each
Quantity
Catalog number: 043179.14
also known as 043179-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16962-47-3
Molecular Formula
F6GeH16N2
InChI Key
LHKQHMLGVWLMTF-UHFFFAOYSA-N
SMILES
[NH4+].[NH4+].F.F.F.F.[F-].[F-].[GeH4]
Molecular Weight (g/mol)
230.76
Total Metal Impurities
0.01% max.

Description

Ammonium hexafluorogermanate is used to substitutes Al with Ge in Y type zeolites.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text