Chromium(III) nitrate nonahydrate, 99.99% min (metals basis), Thermo Scientific Chemicals
Chromium(III) nitrate nonahydrate, 99.99% min (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chromium(III) nitrate nonahydrate, 99.99% min (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number 043182.22
also known as 043182-22
Price (USD)/ Each
564.65
Online exclusive
627.00 
Save 62.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
564.65
Online exclusive
627.00 
Save 62.35 (10%)
Add to cart
Chromium(III) nitrate nonahydrate, 99.99% min (metals basis), Thermo Scientific Chemicals
Catalog number043182.22
Price (USD)/ Each
564.65
Online exclusive
627.00 
Save 62.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7789-02-8
IUPAC Namechromium(3+) trinitrate
Molecular FormulaCrN3O9
InChI KeyPHFQLYPOURZARY-UHFFFAOYSA-N
SMILES[Cr+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (special)?2.5 % of Theoretical value (Cr)
FormCrystals
Total Metal Impurities≤0.01% (100 ppm)
Appearance (Color)Violet
Chromium nitrate is used in the manufacture of alkali metal-free catalysts and in preservation. It is used in the preparation of chrome catalysts, in textile printing operations, and as a corrosion inhibitor. Chromium(III) nitrate as a source of chromium is added to food supplements for nutritional purposes .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only