Nickel(II) chloride hexahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Nickel(II) chloride hexahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel(II) chloride hexahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 043185.14
also known as 043185-14
Price (USD)/ Each
151.65
Online exclusive
168.00 
Save 16.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
151.65
Online exclusive
168.00 
Save 16.35 (10%)
Add to cart
Nickel(II) chloride hexahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number043185.14
Price (USD)/ Each
151.65
Online exclusive
168.00 
Save 16.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7791-20-0
IUPAC Namenickel(2+) hexahydrate dichloride
Molecular FormulaCl2H12NiO6
InChI KeyLAIZPRYFQUWUBN-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Ni++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green
FormPowder and/or lumps or chunks
Assay from Suppliers CofANi (Assay: ≥24.2 to ≤26.0%)
Total Metal Impurities≤10ppm (≤0.0010%)
Used in the manufacture of all kinds of nickel compounds, preservatives and chemical reaction catalyst. Also used in electroplating, analytical chemistry, used for analysis reagent and in gas masks to absorb NH3. It has also been utilized in environmental studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only