Thermo Scientific Chemicals

Ammonium hexafluorotitanate, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 043192.22
100 g, Each
Thermo Scientific Chemicals

Ammonium hexafluorotitanate, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 043192.22
100 g, Each
Quantity
Catalog number: 043192.22
also known as 043192-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16962-40-6
IUPAC Name
titanium(4+) diammonium hexafluoride
Molecular Formula
F6H8N2Ti
InChI Key
NMGYKLMMQCTUGI-UHFFFAOYSA-J
SMILES
[NH4+].[NH4+].[F-].[F-].[F-].[F-].[F-].[F-].[Ti+4]
Total Metal Impurities
0.01% max.

Description

Ammonium hexafluorotitanate is used as an anti-corrosive cleaning agent. It is also used for the production of ceramics and glass. Further, it is used in the preparation of synthetic gems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text