Strontium nitrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Strontium nitrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Strontium nitrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 043193.14
also known as 043193-14
Price (USD)/ Each
216.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
216.00
Add to cart
Strontium nitrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number043193.14
Price (USD)/ Each
216.00
-
Add to cart
Chemical Identifiers
CAS10042-76-9
IUPAC Namestrontium(2+) dinitrate
Molecular FormulaN2O6Sr
InChI KeyDHEQXMRUPNDRPG-UHFFFAOYSA-N
SMILES[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Used in the manufacture of lights, flares, flame tube, the TV picture tube with the red light emitting agent, red fireworks, optical glass and phosphor. It is used as a colorant (red) in pyrotechnics and is also used as an oxidizer in pyrotechnics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only