Thermo Scientific Chemicals

Magnesium oxide, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog number: 043196.14
25 g, Each
Thermo Scientific Chemicals

Magnesium oxide, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog number: 043196.14
25 g, Each
Quantity
Catalog number: 043196.14
also known as 043196-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1309-48-4
IUPAC Name
oxomagnesium
Molecular Formula
MgO
InChI Key
CPLXHLVBOLITMK-UHFFFAOYSA-N
SMILES
O=[Mg]
Total Metal Impurities
0.05% max.

Description

Magnesium oxide is used as a laboratory reagent, and a good reference white color for colorimetry. Magneisum oxide microspheres is used for enrichment method of polycyclic aromatic hydrocarbons. It is also used in groundwater studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text