Thermo Scientific Chemicals

Zirconium dinitrate oxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 043224.14
25 g, Each
Thermo Scientific Chemicals

Zirconium dinitrate oxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 043224.14
25 g, Each
Quantity
Catalog number: 043224.14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
14985-18-3
IUPAC Name
(nitrooxy)(oxo)zirconio nitrate
Molecular Formula
N2O7Zr
InChI Key
JWBWFTANBHYHBC-UHFFFAOYSA-N
SMILES
[O-][N+](=O)O[Zr](=O)O[N+]([O-])=O
Appearance (Color)
White
Form
Powder or crystals or crystalline powder
Total Metal Impurities
0.1% max.

Description

Zirconium dinitrate oxide hydrate is used as a precursor in the production of Pb(Zr0.50Ti0.50)O3 ferroelectric thin films and silicaand alumina-doped nanocrystalline ceria/zirconia. It is also used to prepare cerium oxide/zirconium oxide mixed oxide catalyst supports.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text