Poly(methyl-3,3,3-trifluoropropylsiloxane), M.W. 14,000, Thermo Scientific Chemicals
Poly(methyl-3,3,3-trifluoropropylsiloxane), M.W. 14,000, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Poly(methyl-3,3,3-trifluoropropylsiloxane), M.W. 14,000, Thermo Scientific Chemicals

Fluorosilicone lubricant
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 043230.14
also known as 043230-14
Price (USD)/ Each
49.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
58.00 
Save 8.35 (14%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
49.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
58.00 
Save 8.35 (14%)
Add to cart
Poly(methyl-3,3,3-trifluoropropylsiloxane), M.W. 14,000, Thermo Scientific Chemicals
Catalog number043230.14
Price (USD)/ Each
49.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
58.00 
Save 8.35 (14%)
-
Add to cart
Chemical Identifiers
CAS63148-56-1
IUPAC Namemethyl(3,3,3-trifluoropropyl)silanone
Molecular FormulaC4H7F3OSi
InChI KeyCUMCLKHSZZGALD-UHFFFAOYSA-N
SMILESC[Si](=O)CCC(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
FormLiquid
Viscosity9,000-11,000 cSt
Fluorosilicone lubricant, lubricant additive, release agent, defoaming Agent, additive to improve chemical resistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only