Thermo Scientific Chemicals

Poly(methyl-3,3,3-trifluoropropylsiloxane), M.W. 14,000, Thermo Scientific Chemicals

Catalog number: 043230.22
100 g, Each
Thermo Scientific Chemicals

Poly(methyl-3,3,3-trifluoropropylsiloxane), M.W. 14,000, Thermo Scientific Chemicals

Catalog number: 043230.22
100 g, Each
Quantity
Catalog number: 043230.22
also known as 043230-22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
63148-56-1
IUPAC Name
methyl(3,3,3-trifluoropropyl)silanone
Molecular Formula
C4H7F3OSi
InChI Key
CUMCLKHSZZGALD-UHFFFAOYSA-N
SMILES
C[Si](=O)CCC(F)(F)F
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
Viscosity
9,000-11,000 cSt
Form
Liquid

Description

Fluorosilicone lubricant, lubricant additive, release agent, defoaming Agent, additive to improve chemical resistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text