Thermo Scientific Chemicals

Tris(dimethylamino)aluminum dimer, Thermo Scientific Chemicals

Catalog number: 043237.03
1 g, Each
Thermo Scientific Chemicals

Tris(dimethylamino)aluminum dimer, Thermo Scientific Chemicals

Catalog number: 043237.03
1 g, Each
Quantity
Catalog number: 043237.03
also known as 043237-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
32093-39-3
IUPAC Name
bis([bis(dimethylamino)alumanyl]dimethylamine)
Molecular Formula
C12H36Al2N6
InChI Key
JGZUJELGSMSOID-UHFFFAOYSA-N
SMILES
CN(C)[Al](N(C)C)N(C)C.CN(C)[Al](N(C)C)N(C)C
Identification (FTIR)
Conforms
Assay by NMR
Conforms
Appearance (Color)
White to yellow to orange
Form
crystalline powder

Description

Preparation of aluminum nitrides

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text