Thermo Scientific Chemicals

Triethylaluminum, 94%, Thermo Scientific Chemicals

Catalog number: 043243.22
100 g, Each
Thermo Scientific Chemicals

Triethylaluminum, 94%, Thermo Scientific Chemicals

Catalog number: 043243.22
100 g, Each
Quantity
Catalog number: 043243.22
also known as 043243-22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
97-93-8
IUPAC Name
triethylalumane
Molecular Formula
C6H15Al
InChI Key
VOITXYVAKOUIBA-UHFFFAOYSA-N
SMILES
CC[Al](CC)CC
Form
Liquid
Assay from Supplier's CofA
≥92.0%

Description

Triethylaluminum is used as a co-catalyst in the industrial production of polyethylene and for the production of medium chain alcohols. Used as a catalyst in Ziegler-Natta polymerization process for vinyl, olefin, diene polymerizations and linear oligomerization and cyclization of unsaturated hydrocarbons. It is also used as a catalyst to produce ethylene gas, chain growth of ethylene, longer chain aluminum alkyls, and in plating aluminum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text