Thermo Scientific Chemicals
Dimethylsuberimidate dihydrochloride, Thermo Scientific Chemicals
Catalog number: 043244.09
10 g, Each
Thermo Scientific Chemicals
Dimethylsuberimidate dihydrochloride, Thermo Scientific Chemicals
Catalog number: 043244.09
10 g, Each
Quantity
Catalog number: 043244.09
also known as 043244-09
Price (USD)
Quantity
-
Have Questions?
Description
Dimethylsuberimidate dihydrochloride is used as a homobifunctional cross-linking reagent for amino groups. It is also used for the modification of tyrosinase to produce biocatalytic enzymatic flow-through detectors. It serves as a cross-linker for preparation of collagen-calcium phosphate composites. Further, it is used in the treatment of avian myeloblastosis virus.